Petroff-Hausser Chambers, Electron Microscopy Sciences | Avantor
Petroff-Hausser Chambers, Electron Microscopy Sciences
Petroff-Hausser Chambers, Electron Microscopy Sciences
Certificates
Petroff-Hausser Chambers, Electron Microscopy Sciences
Petroff-Hausser Chambers, Electron Microscopy Sciences
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
New improved Neubauer ruling pattern with 1/400 sq. mm of ruling. Designed for the counting of bacteria, sperm, blood platelets, and the content of vaccines.
Cell is 0.02 mm deep with 1.5 mm thick glass slide allows the use off dark field microscopy.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Petroff Hausser chamber, no grid with case (3900sp)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,327.13
    $2,327.13
    Each
       $2,327.13
    Supplier Number: 63512-22
  • Petroff Hausser chamber 10 (3901)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,228.08
    $3,228.08
    Each
       $3,228.08
    Supplier Number: 63512-23
  • Petroff Hausser chamber 20 (3902)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,944.52
    $2,944.52
    Each
       $2,944.52
    Supplier Number: 63512-24
  • Petroff Hausser chamber, 10 µm without grid
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,944.52
    $2,944.52
    Each
       $2,944.52
    Supplier Number: 63512-25
  • Petroff Hausser chamber (3900)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,327.13
    $2,327.13
    Each
       $2,327.13
    Supplier Number: 63512-20