Wash Bottles, Permanently Labeled/Color-Coded, Electron Microscopy Sciences | Avantor
Wash Bottles, Permanently Labeled/Color-Coded, Electron Microscopy Sciences
Wash Bottles, Permanently Labeled/Color-Coded, Electron Microscopy Sciences
Certificates
Wash Bottles, Permanently Labeled/Color-Coded, Electron Microscopy Sciences
Wash Bottles, Permanently Labeled/Color-Coded, Electron Microscopy Sciences
8 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Permanently labeled/color-coded wash bottles.
Pack of six - Variety includes: Acetone, Methanol, Water, Iso-propyl Alcohol, Ethanol and Toluene. Toluene Wash Bottle is for short term storage only.
These safe wash bottles, which are permanently labeled and color-coded, eliminate the risk of dispensing the wrong liquid by mistake. All of these wide-mouth (easy transfer liquid) polyethylene bottles have polyalomer caps and a fine-tip-nozzle that may be cut back for heavier flow. Capacity is 16 oz. (500 ml), low density polyethylene.
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Availability
    Price
  • Ethyl Acetate Wash Bottle
    500 ml (16.9 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $32.73
    $32.73
    Each
       $32.73
    Supplier Number: 64155
  • Dichloromethane Wash Bottle
    500 ml (16.9 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $35.57
    $35.57
    Each
       $35.57
    Supplier Number: 64153
  • Isopropyl Alcohol Wash Bottle
    500 ml (16.9 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $35.54
    $35.54
    Each
       $35.54
    Supplier Number: 64157
  • Water Wash Bottle
    500 ml (16.9 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $33.58
    $33.58
    Each
       $33.58
    Supplier Number: 64165
  • Soap Wash Bottle
    500 ml (16.9 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $33.73
    $33.73
    Each
       $33.73
    Supplier Number: 64161
  • Ethanol Wash Bottle
    500 ml (16.9 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $35.62
    $35.62
    Each
       $35.62
    Supplier Number: 64154
  • Toluene Wash Bottle
    500 ml (16.9 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $33.73
    $33.73
    Each
       $33.73
    Supplier Number: 64162
  • Acetonitrile Wash Bottle
    500 ml (16.9 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $42.84
    $42.84
    Each
       $42.84
    Supplier Number: 64151