Perfect Loop, Electron Microscopy Sciences
The Perfect loop is used to place your thin sections that have been cut on the ultramicrotome, easily on the grid mesh without creases.
The Perfect loop for light microscopy has outside diameter of 7 mm which is suitable for large sections.
The Perfect loop is used to place your thin sections that have been cut on the ultramicrotome, easily on the grid mesh without creases.
The Perfect loop for light microscopy has outside diameter of 7 mm which is suitable for large sections.
Product Family Options
- Désignation
- : 70944
- : 70945
- Perfect Loop OnlyEach$1,355.35Pack of 5$1,033.11: 70946
- : 70948
- : 70940
- : 70941
- Light Microscopy Perfect Loop OnlyEach$1,470.60Pack of 5$892.73: 70942
- Light Microscopy Loop Handle-: 70943