Parasite Magnifier, Electron Microscopy Sciences
100491-434
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Magnification:2.5×, 5×
- Description:Parasite Magnifier
- Cat. no.:100491-434
- Supplier No.:68110
An improved version of the conventional magnifying glass.
A 20 mm diameter parasite-like lens is cemented on a large 75 mm diameter lens. Magnification of the large one is 2.5X, while the small lens area has the magnification of 5X. The magnified image is very sharp and clear. Weight: 111g