Parafilm M, Electron Microscopy Sciences
Parafilm M, Electron Microscopy Sciences
Certificates
Parafilm M, Electron Microscopy Sciences
Parafilm M, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Laboratory film is used to prevent evaporation, contamination and hydration. Seals labware quickly and efficiently. It is ideal for electron microscopy drop staining and to pick up thin films when coating grids.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Largeur
    Availability
    Price
  • Parafilm M, 1" core
    76.2 m (250')
    2" (50 mm)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $118.17
    $118.17
    Each
       $118.17
    Supplier Number: 70991
  • Parafilm M, 1" core
    38.1 m (125')
    4" (101 mm)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $196.74
    $196.74
    Each
       $196.74
    Supplier Number: 70990