About this item
New and improved staining guard pen.
New and improved staining guard pen. The Super PAP Pen outperforms the original PAP Pen. The Super PAP Pen may be used at temperatures as high as 120°F. (The old style PAP Pen worked only up to 90°F). With this high temperature feature, one can now use the Super PAP Pen to perform the peroxidase-anti-peroxidase method, ABC method, and Fluorescent antibodies method with satisfactory results.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- UN No.:1993
- Boiling point:71 °C (160 °F)