Needle Probe Set, Electron Microscopy Sciences | Avantor
Needle Probe Set, Electron Microscopy Sciences
Needle Probe Set, Electron Microscopy Sciences
Certificates
Needle Probe Set, Electron Microscopy Sciences
Needle Probe Set, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Set consists of one lightweight precision handle and four interchangeable tools. Can be used for manipulating micro samples, scribing, spring assembly, and probing. The handle is finely balanced, non-rolling gold anodized with split collet and a knurled lock nut. The length is 120 mm (4.7"). Precision forged and ground tools are hardened and nickel plated tool steel with sharp tips.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • Replacement Probe #3
    120 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $15.51
    $15.51
    Each
       $15.51
    Supplier Number: 62130-03
  • Replacement Probe #4
    120 mm
    Discontinued
    Supplier Number: -