Tripod Polisher® TEM/SEM Sample Preparation, 590, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
A Tripod Polisher® was designed to prepare accurately micro sizes of TEM and SEM samples.
- Limited ion milling
- No strong chemicals are used to prepare the specimen
- Reduce preferential thinning, radiation damage, and heating of the sample
Used to prepare both plan-view and cross-sections from a variety of sample materials, such as ceramics, composites, metals and, geological specimens. The same sample can be studied with both SEM and TEM and hence reduces sample preparation.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together