Mini-Tool Sets, Electron Microscopy Sciences
À propos de cet article
Mini Tool Set A: 5 piece set includes scriber, chisel, knife, triangular scraper and oval graver - each razor sharp. Hexagonal anodized aluminum handles, 140 mm (5.5") long. Hardened tool steel tips, 1.5 mm (1/16") diameter, are easily sharpened or pointed.
Mini Tool Set B: Contains eight tool tips with integrated, color-coded handles which are light-weight, non rolling and well balanced. Overall tool length is approximately 127 mm (5"). Tip size is 0.8 mm (.030"). Tip material is hardened tool steel, nickel plated. Set contains: Straight Micro-Prober, 30° Micro-Prober, 90° Micro-Prober, Micro-Chisel, Micro-Knife, Micro-Graver, Micro-Diamond Scriber and, Micro-Measuring Scale.
Mini Tool Set C: Consists of three different tools: Straight Scriber with sharp tip, Sharp Triangular Scraper, Bent Prober with sharp tip - all with integrated hexagonal aluminum handles. Tip material is hardened steel, nickel plated. Tip size is 0.8 mm by 25 mm long. Overall length of tools is approximately 140 mm (5.5").
Delivery information: Comes with a protective plastic case.
1 Options Available Below
Product Details & Documents
Mini Tool Set A: 5 piece set includes scriber, chisel, knife, triangular scraper and oval graver - each razor sharp. Hexagonal anodized aluminum handles, 140 mm (5.5") long. Hardened tool steel tips, 1.5 mm (1/16") diameter, are easily sharpened or pointed.
Mini Tool Set B: Contains eight tool tips with integrated, color-coded handles which are light-weight, non rolling and well balanced. Overall tool length is approximately 127 mm (5"). Tip size is 0.8 mm (.030"). Tip material is hardened tool steel, nickel plated. Set contains: Straight Micro-Prober, 30° Micro-Prober, 90° Micro-Prober, Micro-Chisel, Micro-Knife, Micro-Graver, Micro-Diamond Scriber and, Micro-Measuring Scale.
Mini Tool Set C: Consists of three different tools: Straight Scriber with sharp tip, Sharp Triangular Scraper, Bent Prober with sharp tip - all with integrated hexagonal aluminum handles. Tip material is hardened steel, nickel plated. Tip size is 0.8 mm by 25 mm long. Overall length of tools is approximately 140 mm (5.5").
Delivery information: Comes with a protective plastic case.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 62097-10
- Mini Tool Set BDiscontinued-Supplier Number: 62097-20
- Replacement KnifeDiscontinued-Supplier Number: 62097-13
- Replacement Oval GraverDiscontinued-Supplier Number: 62097-15