Miniature Measuring Scales, Electron Microscopy Sciences
Miniature Measuring Scales, Electron Microscopy Sciences
Scale #3, calibrated to 0.1 mm, 10 mm total measurement, 83 mm (3¼")
Catalog # 100503-066
Supplier #:  62135-03
Miniature Measuring Scales, Electron Microscopy Sciences
Catalog # 100503-066
Miniature Measuring Scales, Electron Microscopy Sciences
Miniature Measuring Scales, Electron Microscopy Sciences
Scale #3, calibrated to 0.1 mm, 10 mm total measurement, 83 mm (3¼")
Price
$196.88
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These unit measuring scales may be used directly under a microscope or magnifier for quick measuring of small specimens, parts, grooves, slots, line widths, holes, chambers, and more.
Precision fabricated from stainless steel, the scales are welded to bendable stainless steel shafts and mounted in color coded, non-rolling handles.
 : Each scale is individually packed in a plastic tube.
Specifications
  • Désignation:
    Scale #3, calibrated to 0.1 mm, 10 mm total measurement
  • Longueur:
    83 mm (3¼")