Mini Vise, Electron Microscopy Sciences
Mini Vise, Electron Microscopy Sciences
Mini vise, 1" jaws
Catalog # 100497-552
Supplier #:  62066-00
Mini Vise, Electron Microscopy Sciences
Catalog # 100497-552
Mini Vise, Electron Microscopy Sciences
Mini Vise, Electron Microscopy Sciences
Mini vise, 1" jaws
Price
$30.90
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The vise is made from cast iron with a heavy red enamel coating.
  • Weight: 13 oz.
This is the smallest vise ever. It comes complete with a clamp that allows for either table or bench-top attachment.
Specifications
  • Désignation:
    Mini vise, 1" jaws