Mini Vacuum Desiccator, Electron Microscopy Sciences
Product Details & Documents
The top part of the desiccator is clear, molded from polycarbonate and see-through from any angle. A polyethylene stopcock with a "PTFE" plug allows for gentle releasing of the vacuum. A peripheral flange around the polypropylene bottom section fits into and snap locks therefore sealing the two halves against a "O" ring. Vacuum retention rating is 740 mm Hg (29"). Inside dimensions are 102mm (4") diameter x 59mm (2 7/16")high. The base is 175mm (6 7/8") diameter.
- Ideal for Small Samples
- Degassing Embedding mixtures
Spécifications
- Désignation:Mini Vacuum Desiccator