Mini-Cleaner Vacuum, Electron Microscopy Sciences
This compact portable vacuum cleaner helps keep your desktop or work surface clean.
- Nozzle gets between computer keyboard keys, card reader slots and other hard to reach crevices
- Strong suction power collects dust, food-crumbs and even staples with ease
Keyboard rubber attachment is used to vacuum the dust and other particles trapped underneath the keyboard keys. Brush and suction tube attachment is used for general cleaning of desktop, air vents, monitor, and computer areas
: Batteries not included.
: This product is not a toy. Small parts could be a choking hazard. Not for children under 5 years. Do not immerse in water. Do not use to vacuum water. Do not use rechargeable batteries - alkaline batteries are recommended. When vacuum will not be used for a long period of time, remove batteries from battery compartment. Batteries may occasionally leak and damage unit, if left inside battery compartment. Store in cool or room temperature areas only. Keep away from direct heat or sunlight.
- Désignation:Mini-cleaner vacuum
- Cat. No.:100491-296
- W×D×H:7×1.3×1.5"
- Power:2 AA batteries
- Weight:3 oz.