Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Description:Microwave calibration slides set
- Cat. no.:100495-468
- Supplier No.:97031-01
Spécifications
À propos de cet article
A calibration slide for microwave staining.
A set which includes two glass slides. Slide #1 has liquid crystal squares corresponding to the following temperatures: 35, 40 and 45 °C; Slide #2 has liquid crystal squares corresponding to the following temperatures: 50, 55 and 60 °C. This calibration set will ensure reproducible programming of the oven to achieve an ideal target temperature of large staining volumes (50 to 200 mL solution).