SuperFrost® PLUS Adhesion Slides, Electron Microscopy Sciences
À propos de cet article
Superfrost Plus(+) glass slide are made by a process that place a permanent positive charge in the microscope slide. The slides electrostatically attract frozen tissue sections and cytology preparations, binding them to the slide and forms a bridge so that covalent bonds develop between formalin fixed sections and the glass.
- Tissue sections and cytological preparations adhere without special adhesives or protein coatings
- Cytological preparations placed on Plus glass resist cell loss during enzyme digestion, DNA denaturation and RNA hybridization
- A higher degree of enzyme digestion can be performed on plus glass without the loss of tissue sections
- Plus glass improves tissue adherence in microwave staining
- Chromic acid washes are redundant for molecular hybridization because the slides are RNAse-free
Plus glass puts an end to background staining. The slides eliminate blue or red background staining which is often encountered in hematoxylin and eosin staining of tissue sections mounted on albuminized glass. No brown background staining occurs when immuno-peroxidase or in situ DNA procedures are performed on our Plus glass.
The slides measure: 25 x 75 mm and are 1 mm thick. Plus slides are Color coded to help keep track of specimens during processing as well as filing.
10 Options Available Below
Product Details & Documents
Superfrost Plus(+) glass slide are made by a process that place a permanent positive charge in the microscope slide. The slides electrostatically attract frozen tissue sections and cytology preparations, binding them to the slide and forms a bridge so that covalent bonds develop between formalin fixed sections and the glass.
- Tissue sections and cytological preparations adhere without special adhesives or protein coatings
- Cytological preparations placed on Plus glass resist cell loss during enzyme digestion, DNA denaturation and RNA hybridization
- A higher degree of enzyme digestion can be performed on plus glass without the loss of tissue sections
- Plus glass improves tissue adherence in microwave staining
- Chromic acid washes are redundant for molecular hybridization because the slides are RNAse-free
Plus glass puts an end to background staining. The slides eliminate blue or red background staining which is often encountered in hematoxylin and eosin staining of tissue sections mounted on albuminized glass. No brown background staining occurs when immuno-peroxidase or in situ DNA procedures are performed on our Plus glass.
The slides measure: 25 x 75 mm and are 1 mm thick. Plus slides are Color coded to help keep track of specimens during processing as well as filing.
Product Family Options
Informations sur le produit
- DésignationColorAvailabilityPrice
- PLUS Adhesion SlideBlueEach$251.05Pack of 144$229.76Supplier Number: 71869-20
- Supplier Number: 71869-21
- PLUS Adhesion SlideGreenEach$251.05Pack of 144$229.76Supplier Number: 71869-30
- Supplier Number: 71869-31
- PLUS Adhesion SlidePinkEach$249.71Pack of 144$226.54Supplier Number: 71869-40
- Supplier Number: 71869-41
- PLUS Adhesion SlideWhiteEach$242.51Pack of 144$220.08Supplier Number: 71869-10
- PLUS Adhesion SlideWhiteCase of 10$2,133.86Each$2,133.86Supplier Number: 71869-11
- PLUS Adhesion SlideYellowEach$247.51Pack of 144$226.54Supplier Number: 71869-50
- Supplier Number: 71869-51