Microscope Slide Folder, Electron Microscopy Sciences
Made from heat resistant injection molded plastic. The Slide Folder™ will hold up to twenty standard microscope slides 75 x 25 mm (3x1").
The Slide Folder™ is made of two parts: a base holding the slides horizontally offering numbered space for easy identification, and transparent doors which can either cover the slides or be swung behind the SlideFolder™ for space conservation while using the tray. The base is available in 5 different colors, offering color-coding for each job. Each slide location is identified with a number from 1 to 20. Removal of the slides is made easy by simply pressing on one end, which will lift the other end up. The two transparent hinged covers offer a full view of each stored slide, even bar codes can be read without removing of the slide. All units are stackable and will resist temperatures between –80°C and +100°C. They are not autoclavable.
Made from heat resistant injection molded plastic. The Slide Folder™ will hold up to twenty standard microscope slides 75 x 25 mm (3x1").
The Slide Folder™ is made of two parts: a base holding the slides horizontally offering numbered space for easy identification, and transparent doors which can either cover the slides or be swung behind the SlideFolder™ for space conservation while using the tray. The base is available in 5 different colors, offering color-coding for each job. Each slide location is identified with a number from 1 to 20. Removal of the slides is made easy by simply pressing on one end, which will lift the other end up. The two transparent hinged covers offer a full view of each stored slide, even bar codes can be read without removing of the slide. All units are stackable and will resist temperatures between –80°C and +100°C. They are not autoclavable.
Product Family Options
- Color
- : 71502-B1
- : 71502-G1
- : 71502-GR1
- : 71502-W10
- : 71502-Y1
- AssortedEach$303.33Pack of 10$275.19: 71502-10
- : 71502-B10
- : 71502-GR10
- YellowEach$336.97Pack of 10$305.71: 71502-Y10
- : 71502-G10
- : 71502-W1
- : 71502-00