MicroPoint™ Uniband Scissors and Featherlite Suture Scissors, Electron Microscopy Sciences
MicroPoint™ Uniband Scissors and Featherlite Suture Scissors, Electron Microscopy Sciences
Certificates
MicroPoint™ Uniband Scissors and Featherlite Suture Scissors, Electron Microscopy Sciences
MicroPoint™ Uniband Scissors and Featherlite Suture Scissors, Electron Microscopy Sciences
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A new concept in scissor technology
  • They have three times the shearing angle to reduce tissue crushing
  • They last far longer than ordinary scissors
  • O.R. Castro-Viejo quality at a floor-grade price
  • They offer consistent cutting at the tip
  • The blades cannot work loose
  • The contact between the blades remains constant
  • Ambidextrous: perfect in right and left hands
  • Easy to pickup and use, no clumsy rings
  • Adjustable shearing contact
  • Lifetime Guarantee: All MicroPoint™ Scissors are guaranteed for life against rust, breakage and rivet loosening
These scissors are available in two configurations: Uniband® and FeatherLite®. The MicroPoint Unibands is 5 inches long, one inch wide and half-an-inch tall.
MicroPoint™ scissors can be used for suture or tissue, and they are manufactured without rings which eliminates the problems with right handed or left handed users. Scissors do not cut by cutting. They cut by shearing.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Blade Length
    Blade Type
    Availability
    Price
  • Uniband LA-4 Scissors
    125 mm
    12 mm (0.47")
    Sharp/Sharp, straight, Smooth/smooth, micro
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $173.29
    $173.29
    Each
       $173.29
    Supplier Number: 77910-25
  • Uniband LA-5 Scissors
    127 mm
    15 mm (0.59")
    Needle nose blades with rounded tips, straight, Smooth/smooth
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $147.59
    $147.59
    Each
       $147.59
    Supplier Number: 77910-30
  • Uniband LA-4C Scissors
    125 mm
    12 mm (0.47")
    Sharp/Sharp, curved, Smooth/smooth
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $175.79
    $175.79
    Each
       $175.79
    Supplier Number: 77910-26
  • Uniband LA-1C Scissors
    125 mm
    20 mm (¾")
    Sharp/sharp, curved blades, All smooth
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $175.79
    $175.79
    Each
       $175.79
    Supplier Number: 77910-11
  • Uniband LA-2C Scissors
    125 mm
    20 mm (¾")
    Sharp/Sharp, curved, Smooth/serrated
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $147.59
    $147.59
    Each
       $147.59
    Supplier Number: 77910-16