Microbeaker Holder, Electron Microscopy Sciences
Microbeaker Holder, Electron Microscopy Sciences
Microbeaker Holder, Electron Microscopy Sciences
Microbeaker Holder, Electron Microscopy Sciences
Brushed aluminum rack holds six 12 mm O.D. microbeakers in unique, stepped "see-through" holes for illuminated and microscopic observation of samples.
Product Family Options
  • Désignation
  • Microbeaker Holder
    Each
    Each - $134.96
    $134.96
    Each
       $134.96
     : 72645
  • Microbeaker Holder
    Each
    Each - $189.71
    $189.71
    Each
       $189.71
     : 72645-10