Membrane Boxes, Electron Microscopy Sciences
Marketsource
Membrane Boxes, Electron Microscopy Sciences
Marketsource
Certificates
Membrane Boxes, Electron Microscopy Sciences
Membrane Boxes, Electron Microscopy Sciences
40 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These unique membrane boxes are for the transfer, storage and shipping of many delicate items. Once your sensitive item is placed in the box the elastic membrane conforms around the enclosed object and protects it from any movement.
  • Protects items from any movement
  • Double elastic membrane on the top and bottom offers security
  • Available in round, rectangular and square shapes
Product Family Options
Product Information
  • Désignation
    Capacité
    Dimensions
    Interior Dimensions
    Availability
    Price
  • Membrane box, #2 round
    54×82 mm
    54 mm ID
    Log in for availability
    Log in for availability
    Each
    Each - $47.62
    $47.62
    Each
       $47.62
    Supplier Number: 64304-20
  • Membrane box, #3 round
    54×82 mm
    54 mm ID
    Log in for availability
    Log in for availability
    Each
    Each - $77.71
    $77.71
    Each
       $77.71
    Supplier Number: 64304-30
  • Membrane box, rectangular
    8 g
    100×50×16 mm
    67×17×9 mm
    Log in for availability
    Log in for availability
    Each
    Each - $69.50
    $69.50
    Each
       $69.50
    Supplier Number: 64304-36
  • Membrane box, rectangular
    100×50×50 mm
    54×17×12 mm
    Log in for availability
    Log in for availability
    Each
    Each - $70.04
    $70.04
    Each
       $70.04
    Supplier Number: 64304-51
    Specifications:
    Marketsource
    Marketsource
  • Membrane box, rectangular
    100×50×67 mm
    20×20 mm
    Log in for availability
    Log in for availability
    Each
    Each - $70.04
    $70.04
    Each
       $70.04
    Supplier Number: 64304-29
    Specifications:
    Marketsource
    Marketsource
  • Membrane box, rectangular
    19 g
    100×75×16 mm
    67×42×9 mm
    Log in for availability
    Log in for availability
    Each
    Each - $95.98
    $95.98
    Each
       $95.98
    Supplier Number: 64304-38
  • Membrane box, rectangular
    31 g
    100×75×25 mm
    62×37×18 mm
    Log in for availability
    Log in for availability
    Each
    Each - $98.80
    $98.80
    Each
       $98.80
    Supplier Number: 64304-39
  • Membrane box, rectangular
    44 g
    125×75×25 mm
    87×37×18 mm
    Log in for availability
    Log in for availability
    Each
    Each - $96.29
    $96.29
    Each
       $96.29
    Supplier Number: 64304-40
  • Membrane box, rectangular
    150×100×25 mm
    92×92×9 mm
    Log in for availability
    Log in for availability
    Each
    Each - $77.85
    $77.85
    Each
       $77.85
    Supplier Number: 64304-52
    Specifications:
    Marketsource
    Marketsource
  • Membrane box, rectangular
    150×100×75 mm
    104×54×59 mm
    Log in for availability
    Log in for availability
    Each
    Each - $79.85
    $79.85
    Each
       $79.85
    Supplier Number: 64304-54
    Specifications:
    Marketsource
    Marketsource
  • Membrane box, rectangular
    132 g
    150×125×25 mm
    112×87×18 mm
    Log in for availability
    Log in for availability
    Each
    Each - $124.73
    $124.73
    Each
       $124.73
    Supplier Number: 64304-42
  • Membrane box, rectangular
    246 g
    150×125×50 mm
    112×79×20 mm
    Log in for availability
    Log in for availability
    Each
    Each - $112.47
    $112.47
    Each
       $112.47
    Supplier Number: 64304-43
  • Membrane box, rectangular
    324 g
    150×150×16 mm
    104×104×40 mm
    Log in for availability
    Log in for availability
    Each
    Each - $111.70
    $111.70
    Each
       $111.70
    Supplier Number: 64304-50
  • Membrane box, rectangular
    56 g
    150×75×25 mm
    112×37×18 mm
    Log in for availability
    Log in for availability
    Each
    Each - $92.03
    $92.03
    Each
       $92.03
    Supplier Number: 64304-41
  • Membrane box, rectangular
    175×100×25 mm
    137×62×18 mm
    Log in for availability
    Log in for availability
    Each
    Each - $87.93
    $87.93
    Each
       $87.93
    Supplier Number: 64304-57
    Specifications:
    Marketsource
    Marketsource
  • Membrane box, rectangular
    175×100×75 mm
    129×54×40 mm
    Log in for availability
    Log in for availability
    Each
    Each - $94.50
    $94.50
    Each
       $94.50
    Supplier Number: 64304-58
    Specifications:
    Marketsource
    Marketsource
  • Membrane box, rectangular
    200×150×50 mm
    154×104×40 mm
    Log in for availability
    Log in for availability
    Each
    Each - $113.08
    $113.08
    Each
       $113.08
    Supplier Number: 64304-59
    Specifications:
    Marketsource
    Marketsource
  • Membrane box, rectangular
    1557 g
    250×200×100 mm
    188×138×60 mm
    Log in for availability
    Log in for availability
    Each
    Each - $261.91
    $261.91
    Each
       $261.91
    Supplier Number: 64304-44
  • Membrane box, rectangular
    3371 g
    250×200×200 mm
    157×135 mm
    Log in for availability
    Log in for availability
    Each
    Each - $224.83
    $224.83
    Each
       $224.83
    Supplier Number: 64304-45
  • Membrane box, rectangular
    250×200×204 mm
    154×40 mm
    Log in for availability
    Log in for availability
    Each
    Each - $129.31
    $129.31
    Each
       $129.31
    Supplier Number: 64304-28
    Specifications:
    Marketsource
    Marketsource
  • Membrane box, rectangular
    250×200×212 mm
    162×18 mm
    Log in for availability
    Log in for availability
    Each
    Each - $131.96
    $131.96
    Each
       $131.96
    Supplier Number: 64304-27
    Specifications:
    Marketsource
    Marketsource
  • Membrane box, rectangular
    211 g
    300×150×16 mm
    267×117×9 mm
    Log in for availability
    Log in for availability
    Each
    Each - $158.53
    $158.53
    Each
       $158.53
    Supplier Number: 64304-46
  • Membrane box, rectangular
    300×150×25 mm
    267×117×9 mm
    Log in for availability
    Log in for availability
    Each
    Each - $138.80
    $138.80
    Each
       $138.80
    Supplier Number: 64304-55
    Specifications:
    Marketsource
    Marketsource
  • Membrane box, rectangular
    300×150×75 mm
    246×96×60 mm
    Log in for availability
    Log in for availability
    Each
    Each - $156.20
    $156.20
    Each
       $156.20
    Supplier Number: 64304-56
    Specifications:
    Marketsource
    Marketsource
  • Membrane box, square
    10 g
    38×38×18 mm
    31×31×9 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $140.79
    $140.79
    Log in for availability
    Pack of 12
    Pack of 12 - $106.08
    $106.08
    Each
       $140.79
    Pack of 12
       $106.08
    Supplier Number: 64304-10
  • Membrane box, square
    30 g
    100×100×16 mm
    67×67×9 mm
    Log in for availability
    Log in for availability
    Each
    Each - $97.63
    $97.63
    Each
       $97.63
    Supplier Number: 64304-11
  • Membrane box, square
    52 g
    100×100×25 mm
    62×62×18 mm
    Log in for availability
    Log in for availability
    Each
    Each - $109.63
    $109.63
    Each
       $109.63
    Supplier Number: 64304-12
  • Membrane box, square
    88 g
    100×100×50 mm
    54×54×40 mm
    Log in for availability
    Log in for availability
    Each
    Each - $102.54
    $102.54
    Each
       $102.54
    Supplier Number: 64304-13
  • Membrane box, square
    100×100×75 mm
    Log in for availability
    Log in for availability
    Each
    Each - $78.49
    $78.49
    Each
       $78.49
    Supplier Number: 64304-22
    Specifications:
    Marketsource
    Marketsource
  • Membrane box, square
    57 g
    125×125×16 mm
    92×92×9 mm
    Log in for availability
    Log in for availability
    Each
    Each - $103.20
    $103.20
    Each
       $103.20
    Supplier Number: 64304-14
  • Membrane box, square
    102 g
    125×125×25 mm
    87×87×18 mm
    Log in for availability
    Log in for availability
    Each
    Each - $103.20
    $103.20
    Each
       $103.20
    Supplier Number: 64304-15
  • Membrane box, square
    187 g
    125×125×50 mm
    79×79×40 mm
    Log in for availability
    Log in for availability
    Each
    Each - $118.47
    $118.47
    Each
       $118.47
    Supplier Number: 64304-16
  • Membrane box, square
    125×125×75 mm
    92×92×60 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $107.72
    $107.72
    Log in for availability
    Pack of 1
    Pack of 1 - $80.80
    $80.80
    Each
       $107.72
    Pack of 1
       $80.80
    Supplier Number: 64304-18
    Specifications:
    Marketsource
    Marketsource
  • Membrane box, square
    465 g
    150×150×100 mm
    88×88×80 mm
    Log in for availability
    Log in for availability
    Each
    Each - $154.32
    $154.32
    Each
       $154.32
    Supplier Number: 64304-17
  • Membrane box, square
    150×150×25 mm
    112×112×18 mm
    Log in for availability
    Log in for availability
    Each
    Each - $107.69
    $107.69
    Each
       $107.69
    Supplier Number: 64304-19
    Specifications:
    Marketsource
    Marketsource
  • Membrane box, square
    150×150×75 mm
    104×104×60 mm
    Log in for availability
    Log in for availability
    Each
    Each - $117.80
    $117.80
    Each
       $117.80
    Supplier Number: 64304-21
    Specifications:
    Marketsource
    Marketsource
  • Membrane box, square
    175×175×25 mm
    137×137×18 mm
    Log in for availability
    Log in for availability
    Each
    Each - $122.56
    $122.56
    Each
       $122.56
    Supplier Number: 64304-23
    Specifications:
    Marketsource
    Marketsource
  • Membrane box, square
    175×175×50 mm
    129×129×40 mm
    Log in for availability
    Log in for availability
    Each
    Each - $122.50
    $122.50
    Each
       $122.50
    Supplier Number: 64304-24
    Specifications:
    Marketsource
    Marketsource
  • Membrane box, square
    200×200×25 mm
    162×162×18 mm
    Log in for availability
    Log in for availability
    Each
    Each - $130.99
    $130.99
    Each
       $130.99
    Supplier Number: 64304-25
  • Membrane box, square
    200×200×50 mm
    154×154×40 mm
    Log in for availability
    Log in for availability
    Each
    Each - $131.12
    $131.12
    Each
       $131.12
    Supplier Number: 64304-26
    Specifications:
    Marketsource
    Marketsource