Measuring Magnifiers, Electron Microscopy Sciences
Measuring Magnifiers, Electron Microscopy Sciences
Measuring Magnifiers, Electron Microscopy Sciences
Measuring Magnifiers, Electron Microscopy Sciences
Measuring Magnifiers, Electron Microscopy Sciences
Excellent achromatic lenses with a photo-printed precise scale.
  • Focusing ring
  • Metric & English scale interchangeable
Product Family Options
  • Désignation
    Grossissement
  • Measuring Magnifier Metric Scale
    Each
    Each - $153.64
    $153.64
    Each
       $153.64
     : 68150
  • English Scale for 7x Magnifier
    Each
    Each - $47.20
    $47.20
    Each
       $47.20
     : 68151
  • English Scale Only for 10x Magnifier
    Each
    Each - $47.20
    $47.20
    Each
       $47.20
     : 68155
  • Measuring Magnifier, Metric Scale
    10×
    Each
    Each - $203.36
    $203.36
    Each
       $203.36
     : 68154
  • English Scale Only for 15x Magnifier
    Each
    Each - $61.70
    $61.70
    Each
       $61.70
     : 68159
Accessories
  •  
    Description
    Size
  • 100491-484
    Scale #2, for 10× magnifiers
    35 mm
    Each - $45.50
  •  
    Description
    Size
  • 100491-486
    Scale #3, for 7× magnifiers
    26 mm
    Each - $45.50
  •  
    Description
    Size
  • 100491-488
    Scale #3, for 10× magnifiers
    35 mm
    Each - $45.50
  •  
    Description
    Size
  • 100491-514
    Scale #10, for 7× magnifiers
    26 mm
    Each - $45.50
  •  
    Description
    Size
  • 100491-516
    Scale #10, for 10× magnifiers
    35 mm
    Each - $45.50
  •  
    Description
    Size
  • 100491-518
    Scale #11, for 7× magnifiers
    26 mm
    Each - $45.50
  •  
    Description
    Size
  • 100491-522
    Scale #12, for 7× magnifiers
    26 mm
    Each - $45.50
  •  
    Description
    Size
  • 100491-524
    Scale #12, for 10× magnifiers
    35 mm
    Each - $45.50
  •  
    Description
    Size
  • 100491-490
    Scale #4, for 7× magnifiers
    26 mm
    Each - $45.50
  •  
    Description
    Size
  • 100491-492
    Scale #4, for 10× magnifiers
    35 mm
    Each - $45.50
  •  
    Description
    Size
  • 100491-508
    Scale #8, for 10× magnifiers
    35 mm
    Each - $45.50
  •  
    Description
    Size
  • 100491-512
    Scale #9, for 10× magnifiers
    35 mm
    Each - $45.50