Water/Ice Bath Rack, Electron Microscopy Sciences | Avantor
Water/Ice Bath Rack, Electron Microscopy Sciences
Water/Ice Bath Rack, Electron Microscopy Sciences
Certificates
Water/Ice Bath Rack, Electron Microscopy Sciences
Water/Ice Bath Rack, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Rust-proof anodized aluminum rack – designed to be used in water/ice baths.
  • Removable cover keeps tubes closed while boiling
  • Handle for easy removal from water baths
  • Will not rust
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • 24-place for 1.5/2.0 ml Tubes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $403.19
    $403.19
    Each
       $403.19
    Supplier Number: 72380-20
  • 40 Place Rectangular Mailing Rack for 0.5 ml Tubes
    Discontinued
    Supplier Number: 72380-24