Light Pipe Scintillators, Electron Microscopy Sciences
À propos de cet article
The light pipe is the connection between the scintillator and the phosphomultiplier tube (PMT). It transfers the photons produced by the scintillator for detection by the PMT.
- Scintillator material coated directly on the light pipe which improves the signal
- Re-coating is typically needed twice per year
Light Pipes are available in optical quality quartz and optical grade acrylic. Cambridge reduced-tip models are only available in acrylic.
Delivery information: Includes one Re-coating Certificate. High voltage wire and corona ring are included, JEOL pipes include a flange.
7 Options Available Below
Product Details & Documents
The light pipe is the connection between the scintillator and the phosphomultiplier tube (PMT). It transfers the photons produced by the scintillator for detection by the PMT.
- Scintillator material coated directly on the light pipe which improves the signal
- Re-coating is typically needed twice per year
Light Pipes are available in optical quality quartz and optical grade acrylic. Cambridge reduced-tip models are only available in acrylic.
Delivery information: Includes one Re-coating Certificate. High voltage wire and corona ring are included, JEOL pipes include a flange.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 82000-10
- Cambridge Light Pipe Scintillator, Quartz, Straight, 74 mmLog in pour la disponibilitéEach$1,650.57Supplier Number: 82000-74
- Cambridge Light Pipe Scintillator, Quartz, Straight, 89 mmLog in pour la disponibilitéEach$1,650.57Supplier Number: 82000-89
- Cambridge Light Pipe Scintillator, Quartz, Straight, 104 mmLog in pour la disponibilitéEach$1,650.57Supplier Number: 82000-104
- Cambridge Light Pipe Scintillator, Quartz, Straight, Up To 200 mmLog in pour la disponibilitéEach$1,650.57Supplier Number: 82000-X
- Cambridge Light Pipe Scintillator, Acrylic, Reduced Tip, 100 mmLog in pour la disponibilitéEach$1,917.43Supplier Number: 82001-100
- Cambridge Light Pipe Scintillator, Acrylic, Reduced Tip, 120 mmLog in pour la disponibilitéEach$1,917.43Supplier Number: 82001-120