Leica* Disposable Microtome Blades, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Made from high quality stainless steel. High and low profile disposable blades for use with all Leica microtomes as well as the Cryocut and Cryostat.
- A teflon-fluorocarbon resin-coated edge allows sections to slide off the edge without distortion
- They minimize compression
Low profile blade measures L x W x T: 75.74 x 8.09 x 0.27 mm (2.98 x 0.318 x 0.010”)
High profile blade measures L x W x T: 75.74 x 14.14 x 0.32 mm (2.98 x 0.557 x 0.012”)
Packaging: 50 blades in a dispenser box.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together