Bright-Line Counting Chamber, Electron Microscopy Sciences
For accurate counting of blood cells platelets, dust/yeast cluster and specimen samples. Sharp contrast from rhodium plated on "V-Load". Triple dividing lines clearly delineate counting area for accurate and precise results. Molded from a single piece of thermal and shock resistant glass with a polished chamber and cover glass assuring precise capillary loading and even cell distribution. Fluid volume is 0.1 mm3/mm2 and 0.00025 mm2 per each square.
Delivery information: The complete set includes one counting chamber, two 0.4 mm cover glass, one red cell pipette, one white cell pipette, tubing, mouthpieces, and storage case.
For accurate counting of blood cells platelets, dust/yeast cluster and specimen samples. Sharp contrast from rhodium plated on "V-Load". Triple dividing lines clearly delineate counting area for accurate and precise results. Molded from a single piece of thermal and shock resistant glass with a polished chamber and cover glass assuring precise capillary loading and even cell distribution. Fluid volume is 0.1 mm3/mm2 and 0.00025 mm2 per each square.
Delivery information: The complete set includes one counting chamber, two 0.4 mm cover glass, one red cell pipette, one white cell pipette, tubing, mouthpieces, and storage case.
Product Family Options
- Désignation
- Leica Bright-Line Chamber Set (1483)-: 63514-10
- Leica Bright-Line Counting Chamber with two 0.4 mm Cover GlassEach$1,275.01: 63514-11
- : 63514-12
- Phase Chamber Set without Cover Glass (1475)-: 63514-20
- : 63514-30
- Red Cell Pipette (Thoma) (1456)-: 63514-31
- : 63514-32
- : 63514-34