Large Glass Slides, Electron Microscopy Sciences
À propos de cet article
These slides are precleaned, individually selected and are suitable for large specimen sections, such as, brain tissue.
In addition to our comprehensive collection of glass slides manufactured by ESCO we now carry large glass slides which are available in 75×38 mm and 75×51 mm.
2 Options Available Below
Product Details & Documents
These slides are precleaned, individually selected and are suitable for large specimen sections, such as, brain tissue.
In addition to our comprehensive collection of glass slides manufactured by ESCO we now carry large glass slides which are available in 75×38 mm and 75×51 mm.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- 75x51 mm glass slide, 1.2 mm thickEach$239.37Gross of 144$239.37Supplier Number: 71862-01
- 75x38 mm glass slide, 1 mm thickEach$212.64Gross of 144$212.64Supplier Number: 71860-01