Large Glass Slide Storage Box, Electron Microscopy Sciences
À propos de cet article
100-Slide Capacity:
These storage boxes are available in two sizes.
100-Slide Capacity:
Durable, wood frame, covered with heavy black embossed paper. The hinges and latches are Nickel plated. A numbered plastic insert provides spacing for the handling of the slides. The box accommodates 100 of 3x2" slides. The box measures 24.7 x 19.7 x 5.7 cm (9 3/4 x 7 4/5 x 2 1/4").
25 Slide Capacity:
Made from wood with numbered partitions which correspond to the index card. Very durable and has a detachable lid. The box accommodates 25 of 3x2" slides.
25 Slide Capacity:
Made from wood with numbered partitions which correspond to the index card. Very durable and has a detachable lid. The box accommodates 25 of 3x2" slides.
2 Options Available Below
Product Details & Documents
100-Slide Capacity:
These storage boxes are available in two sizes.
100-Slide Capacity:
Durable, wood frame, covered with heavy black embossed paper. The hinges and latches are Nickel plated. A numbered plastic insert provides spacing for the handling of the slides. The box accommodates 100 of 3x2" slides. The box measures 24.7 x 19.7 x 5.7 cm (9 3/4 x 7 4/5 x 2 1/4").
25 Slide Capacity:
Made from wood with numbered partitions which correspond to the index card. Very durable and has a detachable lid. The box accommodates 25 of 3x2" slides.
25 Slide Capacity:
Made from wood with numbered partitions which correspond to the index card. Very durable and has a detachable lid. The box accommodates 25 of 3x2" slides.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- 100 Microscope Slide Box, 3x2" SlidesDiscontinued-Supplier Number: -
- 25 Microscope Slide Box, 3x2" SlidesEach$1,417.86Pack of 10$1,108.61Supplier Number: 71481-10
- Supplier Number: 71481-01
- Discontinued-Supplier Number: -