Lab-Tek® Chamber Slide™ System, Electron Microscopy Sciences
À propos de cet article
Permanox® is a special molding resin, which meets the growing requirements of most anchorage dependent cells; also excellent chemical resistance allows for cell fixation and the staining of organic solvents for EM and other techniques. Low oxygen permeation makes Permanox® ideal for anoxic cell growth.
- Developed for microscopy of in situ cell culture
- Cells grow on a standard microscope slide
- Media chamber can be removed when culturingis complete
- Useful for viral and mycoplasma testing, chromosome studies toxicity tests and immunocytology
- Permanox® plastic and glass do not interfere with fluorescent labels
The chamber slides combine a media-containing chambered super-structure with a standard soda lime glass slide or Permanox® plastic slide, sealing the two together with a biologically inert silicone gasket. After cells are grown, the chamber and gasket are easily removed. The slide may then be fixed, stained, cover slipped, and filed with the usual convenience. These slides, once fixed and stained, can be embedded using our Slide Duplicating Mold (Cat. 70170). Lab-Tek® Chamber Slide™ System all come packed in a sterile sealed tray, 8 units per tray, 2 trays per pack and 12 trays per case.
20 Options Available Below
Product Details & Documents
Permanox® is a special molding resin, which meets the growing requirements of most anchorage dependent cells; also excellent chemical resistance allows for cell fixation and the staining of organic solvents for EM and other techniques. Low oxygen permeation makes Permanox® ideal for anoxic cell growth.
- Developed for microscopy of in situ cell culture
- Cells grow on a standard microscope slide
- Media chamber can be removed when culturingis complete
- Useful for viral and mycoplasma testing, chromosome studies toxicity tests and immunocytology
- Permanox® plastic and glass do not interfere with fluorescent labels
The chamber slides combine a media-containing chambered super-structure with a standard soda lime glass slide or Permanox® plastic slide, sealing the two together with a biologically inert silicone gasket. After cells are grown, the chamber and gasket are easily removed. The slide may then be fixed, stained, cover slipped, and filed with the usual convenience. These slides, once fixed and stained, can be embedded using our Slide Duplicating Mold (Cat. 70170). Lab-Tek® Chamber Slide™ System all come packed in a sterile sealed tray, 8 units per tray, 2 trays per pack and 12 trays per case.
Product Family Options
Informations sur le produit
- DésignationNo. of TraysCapacitéAvailabilityPrice
- 16-well glass slide6Each$1,719.94Pack of 6$1,299.82Supplier Number: 70421
- Flaskette glass slide2Box of 2$613.79Each$613.79Supplier Number: 70417
- 16-well glass slide2Box of 2$613.79Each$613.79Supplier Number: 70420
- Coverglass 22x74 mm2 oz.Box of 2$212.93Each$212.93Supplier Number: 70423
- Lab Tek glass slide with 1 chamber6Each$1,507.43Pack of 6$1,147.44Supplier Number: 70384
- Permanox® slide with 2 chambers6Each$1,755.31Pack of 6$1,333.13Supplier Number: 70393
- Lab Tek glass slide with 4 chambers12Box of 12$3,044.23Each$3,044.23Supplier Number: 70407
- Lab Tek glass slide with 8 chambers12Box of 8$3,075.97Each$3,075.97Supplier Number: 70415
- Flaskette glass slide6Each$1,719.94Pack of 6$1,299.82Supplier Number: 70418
- Permanox® slide with 4 chambers2Box of 2$672.83Each$672.83Supplier Number: 70400
- Lab Tek glass slide with 1 chamber2Each$461.82Pack of 2$418.98Supplier Number: 70381
- Permanox® slide with 1 chamber6Each$1,729.44Pack of 6$1,314.09Supplier Number: 70383
- Permanox® slide with 4 chambers12Case of 12$3,535.24Each$3,535.24Supplier Number: 70406
- Permanox® slide with 8 chambers12Case of 12$3,619.24Each$3,619.24Supplier Number: 70416
- Permanox® slide with 8 chambers2Box of 2$725.52Each$725.52Supplier Number: 70410
- Permanox® slide with 1 chamber12Discontinued-Supplier Number: 70386
- Flaskette glass slide12Discontinued-Supplier Number: 70419
- Lab Tek glass slide with 2 chambers12Discontinued-Supplier Number: 70397
- Permanox® slide with 2 chambers12Discontinued-Supplier Number: 70396
- Lab Tek glass slide with 4 chambers2Discontinued-Supplier Number: 70401
- 16-well glass slide12Discontinued-Supplier Number: 70422
- Permanox® slide with 2 chambers2Box of 2$625.88Each$625.88Supplier Number: 70390
- Lab Tek glass slide with 2 chambers6Box of 6$1,544.86Each$1,544.86Supplier Number: 70394
- Permanox® slide with 8 chambers6Each$1,508.69Pack of 6$1,368.82Supplier Number: 70413
- Permanox® slide with 1 chamber2Box of 2$622.93Each$622.93Supplier Number: 70380
- Lab Tek glass slide with 2 chambers2Discontinued-Supplier Number: 70391
- Lab Tek glass slide with 8 chambers2Box of 2$582.14Each$582.14Supplier Number: 70411