Adhesive Labels on Laser Sheets, Round, Electron Microscopy Sciences
Adhesive Labels on Laser Sheets, Round, Electron Microscopy Sciences
Adhesive Labels on Laser Sheets, Round, Electron Microscopy Sciences
Adhesive Labels on Laser Sheets, Round, Electron Microscopy Sciences
Tough-Spots® Labels are pre-cut peel-off labels specially for microcentrifuge tube tops. This chemically resistant, flexible labels that can withstand boiling, freezing to −196 °C, sustained humidity.
  • Polyolefin labels resist most organic solvents and caustic agents
  • They adhere to most plastics, glass and metals without cracking, peeling, or degrading
  • Heat-resistant, stay-flat sheets will not jam in laser printers
  • Label sheets are top-coated to readily accept laser printing
  • Assortment includes four sheets of blue, green, red, yellow and orange color
Made of polyolefin label material and acrylic adhesive. This unique combination expands and contracts together with temperature fluctuations resulting in a strong adhesive bond.
Product Family Options
  • Désignation
    Color
    Diameter
    Packaging
  • Adhesive Tough-Spots on laser sheets, round
    Assorted
    3/8"
    3840 labels/pack
    Each
    Each - $209.27
    $209.27
    Pack of 3,840
    Pack of 3,840 - $177.56
    $177.56
    Each
       $209.27
    Pack of 3,840
       $177.56
     : 77585-05
  • Adhesive Tough-Spots on laser sheets, round
    Blue
    3/8"
    3840 labels/pack
    Each
    Each - $218.42
    $218.42
    Pack of 3,840
    Pack of 3,840 - $178.38
    $178.38
    Each
       $218.42
    Pack of 3,840
       $178.38
     : 77585-B
  • Adhesive Tough-Spots on laser sheets, round
    Green
    3/8"
    3840 labels/pack
    Each
    Each - $218.42
    $218.42
    Pack of 3,840
    Pack of 3,840 - $178.38
    $178.38
    Each
       $218.42
    Pack of 3,840
       $178.38
     : 77585-G
  • Adhesive Tough-Spots on laser sheets, round
    Orange
    3/8"
    3840 labels/pack
    Each
    Each - $218.42
    $218.42
    Pack of 3,840
    Pack of 3,840 - $178.38
    $178.38
    Each
       $218.42
    Pack of 3,840
       $178.38
     : 77585-O
  • Adhesive Tough-Spots on laser sheets, round
    Red
    3/8"
    3840 labels/pack
    Each
    Each - $218.42
    $218.42
    Pack of 3,840
    Pack of 3,840 - $178.38
    $178.38
    Each
       $218.42
    Pack of 3,840
       $178.38
     : 77585-R
  • Adhesive Tough-Spots on laser sheets, round
    White
    3/8"
    3840 labels/pack
    Each
    Each - $210.25
    $210.25
    Pack of 3,840
    Pack of 3,840 - $178.38
    $178.38
    Each
       $210.25
    Pack of 3,840
       $178.38
     : 77585-W
  • Adhesive Tough-Spots on laser sheets, round
    Yellow
    3/8"
    3840 labels/pack
    Each
    Each - $240.60
    $240.60
    Pack of 3,840
    Pack of 3,840 - $275.50
    $275.50
    Each
       $240.60
    Pack of 3,840
       $275.50
     : 77585-Y
  • Adhesive laser labels, Tough-Spots, round
    White
    7/16"
    4680 labels/pack
    Each
    Each - $217.44
    $217.44
    Pack of 4,680
    Pack of 4,680 - $177.56
    $177.56
    Each
       $217.44
    Pack of 4,680
       $177.56
     : 77585-44
  • Adhesive laser labels, Tough-Spots, round
    Assorted
    1/2"
    3840 labels/pack
    Each
    Each - $217.44
    $217.44
    Pack of 3,840
    Pack of 3,840 - $177.56
    $177.56
    Each
       $217.44
    Pack of 3,840
       $177.56
     : 77587-05
  • Adhesive laser labels, Tough-Spots, round
    Blue
    1/2"
    3840 labels/pack
    Each
    Each - $218.42
    $218.42
    Pack of 3,840
    Pack of 3,840 - $178.38
    $178.38
    Each
       $218.42
    Pack of 3,840
       $178.38
     : 77587-B
  • Adhesive laser labels, Tough-Spots, round
    Green
    1/2"
    3840 labels/pack
    Each
    Each - $218.42
    $218.42
    Pack of 3,840
    Pack of 3,840 - $178.38
    $178.38
    Each
       $218.42
    Pack of 3,840
       $178.38
     : 77587-G
  • Adhesive laser labels, Tough-Spots, round
    Orange
    1/2"
    3840 labels/pack
    Each
    Each - $218.42
    $218.42
    Pack of 3,840
    Pack of 3,840 - $178.38
    $178.38
    Each
       $218.42
    Pack of 3,840
       $178.38
     : 77587-O
  • Adhesive laser labels, Tough-Spots, round
    Red
    1/2"
    3840 labels/pack
    Each
    Each - $218.42
    $218.42
    Pack of 3,840
    Pack of 3,840 - $178.38
    $178.38
    Each
       $218.42
    Pack of 3,840
       $178.38
     : 77587-R
  • Adhesive laser labels, Tough-Spots, round
    White
    1/2"
    3840 labels/pack
    Each
    Each - $217.44
    $217.44
    Pack of 3,840
    Pack of 3,840 - $177.56
    $177.56
    Each
       $217.44
    Pack of 3,840
       $177.56
     : 77587-W
  • Adhesive laser labels, Tough-Spots, round
    Yellow
    1/2"
    3840 labels/pack
    Each
    Each - $240.60
    $240.60
    Pack of 3,840
    Pack of 3,840 - $275.50
    $275.50
    Each
       $240.60
    Pack of 3,840
       $275.50
     : 77587-Y
  • Adhesive laser labels, Tough-Spots, round
    White
    3/4"
    2160 labels/pack
    Each
    Each - $215.38
    $215.38
    Pack of 2,160
    Pack of 2,160 - $175.89
    $175.89
    Each
       $215.38
    Pack of 2,160
       $175.89
     : 77588-75
  • Adhesive laser labels, Tough-Spots, round
    White
    1"
    1260 labels/pack
     : 77588-100