Premium Abrasive Cut-off Wheels for Sectioning, Electron Microscopy Sciences
À propos de cet article
Silicone Carbide disc are the finest wet or dry abrasive discs for sample preparation
Wheels are available in 9 different grit sizes, ranging from coarse to fine, on plain back or water resistant pressure sensitive adhesive back.
Pressure sensitive adhesive backed discs eliminates the need to transfer to plates. Silicon Carbide Abrasive Cut-Off Wheels offered long life, and affordably priced sectioning. All wheels are 9" (228.6 mm) diameter 0.062” (1.6 mm) thick; 1¼” (31mm) arbor.
Ordering information: Available in 8" diameter discs as a standard and 12" discs available upon request.
10 Options Available Below
Product Details & Documents
Silicone Carbide disc are the finest wet or dry abrasive discs for sample preparation
Wheels are available in 9 different grit sizes, ranging from coarse to fine, on plain back or water resistant pressure sensitive adhesive back.
Pressure sensitive adhesive backed discs eliminates the need to transfer to plates. Silicon Carbide Abrasive Cut-Off Wheels offered long life, and affordably priced sectioning. All wheels are 9" (228.6 mm) diameter 0.062” (1.6 mm) thick; 1¼” (31mm) arbor.
Ordering information: Available in 8" diameter discs as a standard and 12" discs available upon request.
Product Family Options
Informations sur le produit
- DésignationGritBackingColorAvailabilityPrice
- Disque en carbure de silicium, 8"240PlainBox of 25127,65 $Each127,65 $Supplier Number: 50250-05
- Disque en carbure de silicium, 8"800PlainBox of 25112,89 $Each112,89 $Supplier Number: 50250-09
- Disque en carbure de silicium, 8"800PSADiscontinued-Supplier Number: 50255-09
- Disque en carbure de silicium, 8"120PlainBox of 25112,89 $Each112,89 $Supplier Number: 50250-03
- Disque en carbure de silicium, 8"320PlainBox of 25112,89 $Each112,89 $Supplier Number: 50250-06
- Disque en carbure de silicium, 8"400PlainBox of 2594,52 $Each94,52 $Supplier Number: 50250-07
- Disque en carbure de silicium, 8"400PSABox of 25141,70 $Each141,70 $Supplier Number: 50255-07
- Disque en carbure de silicium, 8"1200PSABox of 25130,24 $Each130,24 $Supplier Number: 50255-10
- Disque en carbure de silicium, 8"600PlainBox of 25112,89 $Each112,89 $Supplier Number: 50250-08
- Disque en carbure de silicium, 8"120PSADiscontinued-Supplier Number: 50255-03
- Disque en carbure de silicium, 8"1200PlainBox of 25112,89 $Each112,89 $Supplier Number: 50250-10
- Disque en carbure de silicium, 8"180PSABox of 25141,70 $Each141,70 $Supplier Number: 50255-04