Vacuum Ovens, Electron Microscopy Sciences
Easy to clean multipurpose vacuum ovens with two control configurations and display options. Radiant warm wall heating system provides uniformity. Conserves chamber space for drying, curing, vacuum embedding, and plating applications.
- Operating temperature range from ambient +5 to 220 °C (428 °F)
- Vacuum level displayed on gauge from 0 to 30" Hg (76.2 cm).
- 220 °C maximum temperature
- Exclusive polycarbonate safety shield
- Two removable aluminum shelves
- Silicon door gasket
- 24 months warranty
Model 3606, 3608 and 3618 Vacuum Ovens feature a hydraulic thermostat to control temperature to ±1.5 °C with a uniformity of ±5 °C @ 100 °C; 25" Hg and built-in oven-temperature protection.
Model 3618P Vacuum Ovens feature programmable microprocessor- based P.I.D. temperature control that regulates to within ±0.2 °C with a uniformity of ±2.4 °C @ 100 °C; 25" Hg. The programmable microprocessor- based temperature control on Model 3618P Vacuum Ovens run at a single set point or utilize a 24-step program of ramps and dwells. These vacuum ovens feature independent vacuum and purge needle values.
Model 3606 and 3606-1 have top-mounted independent evacuation and venting vacuum fittings; all other models features front-mounted three-way valve for evacuation, venting, and purging of inert gases (e.g. nitrogen or argon) with vacuum fitting on front of unit.
Easy to clean multipurpose vacuum ovens with two control configurations and display options. Radiant warm wall heating system provides uniformity. Conserves chamber space for drying, curing, vacuum embedding, and plating applications.
- Operating temperature range from ambient +5 to 220 °C (428 °F)
- Vacuum level displayed on gauge from 0 to 30" Hg (76.2 cm).
- 220 °C maximum temperature
- Exclusive polycarbonate safety shield
- Two removable aluminum shelves
- Silicon door gasket
- 24 months warranty
Model 3606, 3608 and 3618 Vacuum Ovens feature a hydraulic thermostat to control temperature to ±1.5 °C with a uniformity of ±5 °C @ 100 °C; 25" Hg and built-in oven-temperature protection.
Model 3618P Vacuum Ovens feature programmable microprocessor- based P.I.D. temperature control that regulates to within ±0.2 °C with a uniformity of ±2.4 °C @ 100 °C; 25" Hg. The programmable microprocessor- based temperature control on Model 3618P Vacuum Ovens run at a single set point or utilize a 24-step program of ramps and dwells. These vacuum ovens feature independent vacuum and purge needle values.
Model 3606 and 3606-1 have top-mounted independent evacuation and venting vacuum fittings; all other models features front-mounted three-way valve for evacuation, venting, and purging of inert gases (e.g. nitrogen or argon) with vacuum fitting on front of unit.
- Catalog No:
- 100498-366
- 100498-370
- 100498-374
- 100501-700
- 100498-360
- 100498-364
- 100498-368
- Operating temperature range:
- 5 to 200 °C
- 5 to 200 °C
- 5 to 200 °C
- 5 to 200 °C
- 5 to 200 °C
- 5 to 200 °C
- 5 to 200 °C
- Vacuum range:
- 0 to 762 mm Hg
- 0 to 762 mm Hg
- 0 to 762 mm Hg
- 0 to 762 mm Hg
- 0 to 762 mm Hg
- 0 to 762 mm Hg
- 0 to 762 mm Hg
Product Family Options
uum Ovens, 120 V
- DésignationCapacitéElectricalInterior DimensionsType
- Oven Model 3608-50.7 cu. ft (19.8 L)120 V10 W x 10 H x 12" DLED ThermometerEach$17,460.54: 63236-10Minority or Woman-Owned Business Enterprise
Specifications
Cat. No.100498-366Operating temperature range5 to 200 °CVacuum range0 to 762 mm HgMinority or Woman-Owned Business Enterprise - Oven Model 36182.3 cu. ft (65 L)120 V14 W x 14 H x 20" DDial ThermometerEach$23,554.36: 63237-10Minority or Woman-Owned Business Enterprise
Specifications
Cat. No.100498-370Operating temperature range5 to 200 °CVacuum range0 to 762 mm HgMinority or Woman-Owned Business Enterprise - Oven Model 3618-52.3 cu. ft (65 L)120 V14 W x 14 H x 20" DLED ThermometerEach$25,055.81: 63238-10Minority or Woman-Owned Business Enterprise
Specifications
Cat. No.100498-374Operating temperature range5 to 200 °CVacuum range0 to 762 mm HgMinority or Woman-Owned Business Enterprise - Oven Model 36060.44 cu. ft (12.5 L)120 V8 W x 8 H x 12" DDial Thermometer-: -Minority or Woman-Owned Business Enterprise
Specifications
Cat. No.100501-700Operating temperature range5 to 200 °CVacuum range0 to 762 mm HgMinority or Woman-Owned Business Enterprise
uum Ovens, 240 V
- DésignationCapacitéElectricalInterior DimensionsType
- Oven Model 36060.44 cu. ft (12.5 L)240 V8 W x 8 H x 12" DDial ThermometerEach$11,775.98: 63234-20Irregular VoltageMinority or Woman-Owned Business Enterprise
Specifications
Cat. No.100498-360Operating temperature range5 to 200 °CVacuum range0 to 762 mm HgIrregular VoltageMinority or Woman-Owned Business Enterprise - Oven Model 3608-10.7 cu. ft (19.8 L)240 V10 W x 10 H x 12" DDial ThermometerEach$12,047.34: 63235-20Irregular VoltageMinority or Woman-Owned Business Enterprise
Specifications
Cat. No.100498-364Operating temperature range5 to 200 °CVacuum range0 to 762 mm HgIrregular VoltageMinority or Woman-Owned Business Enterprise - Oven Model 3608-60.7 cu. ft (19.8 L)240 V10 W x 10 H x 12" DLED ThermometerEach$15,080.52: 63236-20Irregular VoltageMinority or Woman-Owned Business Enterprise
Specifications
Cat. No.100498-368Operating temperature range5 to 200 °CVacuum range0 to 762 mm HgIrregular VoltageMinority or Woman-Owned Business Enterprise