Wooden Picks, Electron Microscopy Sciences
Product Details & Documents
They are round and double tapered; used for cleaning pole pieces and parts of the EM. Also used for orienting the specimen during the embedding stage.
Spécifications
- Désignation:Wood Picks