Gilder Square Mesh, Electron Microscopy Sciences | Avantor
Gilder Square Mesh, Electron Microscopy Sciences
Gilder Square Mesh, Electron Microscopy Sciences
Certificates
Gilder Square Mesh, Electron Microscopy Sciences
Gilder Square Mesh, Electron Microscopy Sciences
8 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Thin Bar Grids have been developed using a new technology to produce ultra-fine grids with thinner cross bars than regular grids.
  • Made of Copper (Cu), Nickel (Ni), Gold (Au)
  • Measures 3.05" in diameter
The result is equally firm specimen support but with 40% more open area for viewing maximum specimen surface area.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Épaisseur
    Bar Size
    Hole Size
    Mesh
    Pitch
    Availability
    Price
  • Square
    Copper
    0.8 mil.
    6 µm
    19 µm
    1000 Mesh
    25 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $118.09
    $118.09
    Each
       $118.09
    Supplier Number: T1000-CU
  • Square
    Nickel
    0.8 mil.
    6 µm
    19 µm
    1000 Mesh
    25 µm
    Discontinued
    Supplier Number: T1000-NI
  • Square
    Copper
    0.8 mil.
    8 µm
    54 µm
    400 Mesh
    62 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $161.43
    $161.43
    Each
       $161.43
    Supplier Number: T400-CU
  • Square
    Gold
    0.8 mil.
    8 µm
    54 µm
    400 Mesh
    62 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $125.81
    $125.81
    Each
       $125.81
    Supplier Number: T400-AU
  • Square
    Nickel
    0.8 mil.
    8 µm
    54 µm
    400 Mesh
    62 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $210.88
    $210.88
    Each
       $210.88
    Supplier Number: T400-NI
  • Square
    Copper
    0.8 mil.
    10 µm
    73 µm
    300 Mesh
    83 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $152.73
    $152.73
    Each
       $152.73
    Supplier Number: T300-CU
  • Square
    Nickel
    0.8 mil.
    10 µm
    73 µm
    300 Mesh
    83 µm
    Discontinued
    Supplier Number: T300-NI
  • Square
    Copper
    0.8 mil.
    12 µm
    113 µm
    200 Mesh
    125 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $141.51
    $141.51
    Each
       $141.51
    Supplier Number: T200-CU
  • Square
    Gold
    0.8 mil.
    12 µm
    113 µm
    200 Mesh
    125 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $88.37
    $88.37
    Each
       $88.37
    Supplier Number: T200-AU
  • Square
    Nickel
    0.8 mil.
    12 µm
    113 µm
    200 Mesh
    125 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $136.83
    $136.83
    Each
       $136.83
    Supplier Number: T200-NI