Square and Grid Scales, Electron Microscopy Sciences
À propos de cet article
Simple grids are convenient for making sketches of the obseved speimen on graph paper.
They are also useful for particle counting.
6 Options Available Below
Product Details & Documents
Simple grids are convenient for making sketches of the obseved speimen on graph paper.
They are also useful for particle counting.
Product Family Options
Informations sur le produit
- DésignationTailleDivisionsAvailabilityPrice
- NE10 Graticule16 mmGrid (net) 0.5 mm pitchDiscontinued-Supplier Number: 68011-16
- Supplier Number: 68015-16
- Supplier Number: 68018-16
- Supplier Number: 68011-19
- Supplier Number: 68015-19
- NE34 Graticule19 mmGrid (net) 0.1 mm squaresDiscontinued-Supplier Number: 68018-19
- Supplier Number: 68011-21
- Supplier Number: 68015-21
- NE34 Graticule21 mmGrid (net) 0.1 mm squaresDiscontinued-Supplier Number: 68018-21