Cryostore® Boxes, Electron Microscopy Sciences
Cryostore® Boxes, Electron Microscopy Sciences
Certificates
Cryostore® Boxes, Electron Microscopy Sciences
Cryostore® Boxes, Electron Microscopy Sciences
8 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from strong Polycarbonate, these durable cryogenic storage boxes are designed to be used at temp-eratures between –190 °C and +121 °C and are auto-clavable
Polycarbonate Boxes for Freezer Storage. Racks stack on top of each other to consolidate storage space. Transparent lid allows for easy viewing of samples and location. Polycarbonate boxes areautoclavable at 120°C for 20 minutes at 15psi. Each box includes drain holes in the base and air vents in the lid to minimize condensation. Available in Blue, Green, Red and Yellow. Removal of vials is facilitated by an innovative tool called the Vial Picker, which is supplied with each storage box.
Made from strong Polycarbonate, these durable cryogenic storage boxes are designed to be used at temp-eratures between –190 °C and +121 °C and are auto-clavable. Different models are available to accommodate ®tubes from 1 to 5 mL.
A transparent cover allows the user to see the contents of the box, and is keyed to the base to prevent misalignment. Being printed with a series of squares (numbered from 1 to 81, or from 1 to 100), the surface accepts writing with markers for better inventory control.
A unique color-coding system uses color plastic grids inserted to separate the cover from the base of the 81-place box. Boxes are made to accept 100 tubes have a colored base instead of grid. A choice of four popular colors is available: blue, green, red and yellow.
A transparent cover allows the user to see the contents of the box, and is keyed to the base to prevent misalignment. Being printed with a series of squares (numbered from 1 to 81, or from 1 to 100), the surface accepts writing with markers for better inventory control.
A unique color-coding system uses color plastic grids inserted to separate the cover from the base of the 81-place box. Boxes are made to accept 100 tubes have a colored base instead of grid. A choice of four popular colors is available: blue, green, red and yellow.
Different models are available to accommodate ®tubes from 1 to 5 mL.
Product Family Options
Informations sur le produit
  • Nb de places
    Color
    For Tubes
    Dimensions
    Availability
    Price
  • 25
    Blue
    1-2 ml Tubes
    3×3×2 1/16"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $154.26
    $154.26
    Log in pour la disponibilité
    Pack of 8
    Pack of 8 - $116.05
    $116.05
    Each
       $154.26
    Pack of 8
       $116.05
    Supplier Number: 61809-B
  • 25
    Red
    1-2 ml Tubes
    3×3×2 1/16"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $204.72
    $204.72
    Log in pour la disponibilité
    Pack of 8
    Pack of 8 - $116.05
    $116.05
    Each
       $204.72
    Pack of 8
       $116.05
    Supplier Number: 61809-R
  • 81
    Blue
    1-2 ml Tubes
    5¼×5¼×2 1/16"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $752.87
    $752.87
    Log in pour la disponibilité
    Each
    Each - $752.87
    $752.87
    Case of 24
       $752.87
    Each
       $752.87
    Supplier Number: 61810-B
  • 81
    Green
    1-2 ml Tubes
    5¼×5¼×2 1/16"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $743.74
    $743.74
    Log in pour la disponibilité
    Each
    Each - $743.74
    $743.74
    Case of 24
       $743.74
    Each
       $743.74
    Supplier Number: 61810-G
  • 81
    Red
    1-2 ml Tubes
    5¼×5¼×2 1/16"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $743.74
    $743.74
    Log in pour la disponibilité
    Each
    Each - $743.74
    $743.74
    Case of 24
       $743.74
    Each
       $743.74
    Supplier Number: 61810-R
  • 81
    Yellow
    1-2 ml Tubes
    5¼×5¼×2 1/16"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $743.74
    $743.74
    Log in pour la disponibilité
    Each
    Each - $743.74
    $743.74
    Case of 24
       $743.74
    Each
       $743.74
    Supplier Number: 61810-Y
  • 81
    Yellow
    5 ml Tubes
    5¼×5¼×3 3/4"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $412.07
    $412.07
    Log in pour la disponibilité
    Each
    Each - $412.07
    $412.07
    Case of 10
       $412.07
    Each
       $412.07
    Supplier Number: 61812-Y
  • 100
    Blue
    1-2 ml Tubes
    5¼×5¼×2 1/16"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $749.96
    $749.96
    Log in pour la disponibilité
    Each
    Each - $749.96
    $749.96
    Case of 24
       $749.96
    Each
       $749.96
    Supplier Number: 61813-B