About this item
A highly porous membrane covers the dish of the desiccant providing good moisture absorption.
These desiccant cartridges can be used in desiccator cabinets or non-vacuum desiccators, analytical balance cases, and precision laboratory instruments. The clear polystyrene dish allows for instant visual inspection of the silica gel color which changes from blue to pale pink as it absorbs moisture.
A highly porous membrane covers the dish of the desiccant providing good moisture absorption. The desiccant can be regenerated by heating cartridges in an oven at 50°C (122°F) overnight. For rapid drying, cut the membrane and transfer the spent gel crystal into a glass or porcelain dish. Then place it in a 150 °C (302 °F) oven for one hour.
The silica gel is inert and non-toxic. Each cartridge is individually sealed in aluminum foil. Dispose of the cartridges after repeated use. Cartridges come in two sizes. Small: 65 mm (2½") diameter. Large: 100 mm (4") diameter.
The silica gel is inert and non-toxic. Each cartridge is individually sealed in aluminum foil. Dispose of the cartridges after repeated use. Cartridges come in two sizes. Small: 65 mm (2½") diameter. Large: 100 mm (4") diameter.
The desiccant can be regenerated by heating cartridges in an oven at 50 °C (122 °F) overnight. For rapid drying, cut the membrane and transfer the spent gel crystal into a glass or porcelain dish. Then place it in a 150 °C (302 °F) oven for one hour.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.