Retrieving Tweezers, Electron Microscopy Sciences
Retrieving Tweezers, Electron Microscopy Sciences
Certificates
Retrieving Tweezers, Electron Microscopy Sciences
Retrieving Tweezers, Electron Microscopy Sciences
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A unique plastic coating on the tips of the tweezers insulates them and provides resistance to heat and chemicals. Ideal when cleaning with ultrasonics.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Availability
    Price
  • Retrieving Tweezers
    6"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $25.49
    $25.49
    Each
       $25.49
    Supplier Number: 62112-06
  • Retrieving Tweezers
    8"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $32.91
    $32.91
    Each
       $32.91
    Supplier Number: 62112-08
  • Retrieving Tweezers
    10"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $41.03
    $41.03
    Each
       $41.03
    Supplier Number: 62112-10
  • Retrieving Tweezers
    12"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $51.68
    $51.68
    Each
       $51.68
    Supplier Number: 62112-12