Replacement Desiccator Plates, Electron Microscopy Sciences | Avantor
Replacement Desiccator Plates, Electron Microscopy Sciences
Replacement Desiccator Plates, Electron Microscopy Sciences
Certificates
Replacement Desiccator Plates, Electron Microscopy Sciences
Replacement Desiccator Plates, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These 4.8 mm thick plates are able to withstand temperatures up to 150°C (300°F) and have excellent resistance to thermal shock, and to chemicals. They are non-combustible and asbestos free. Round plates are perforated with 4 mm (0.165”) diameter holes on 12.7 mm (1/2”) centers in a straight line pattern. They are available in three sizes, and fit our desiccators.
Product Family Options
Informations sur le produit
  • Taille
    Availability
    Price
  • 230 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $456.25
    $456.25
    Each
       $456.25
    Supplier Number: 71242-23
  • 140 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $263.51
    $263.51
    Each
       $263.51
    Supplier Number: 71242-15
  • 190 mm
    Discontinued
    Supplier Number: -