JB-4™ Embedding Kit, Electron Microscopy Sciences
JB-4™ Embedding Kit, Electron Microscopy Sciences
JB-4™ Embedding Kit, Electron Microscopy Sciences
A water soluble embedding media which is based on Glycol Methacrylate (GMA) plastic embedding. It is intended for use in preparing samples for high resolution microscopes (HRLM). The catalyzed monomer acts as a dehydration and infiltration agent. Therefore, complete dehydration through 100% ethanol is not necessary although recommended for large or dense tissues. Conventional paraffin sections have a greater degree of shrinkage and produce inadequate morphology when compared to JB-4.JB-4 Kit Consists of 800 mL Solution A, 30 mL Solution B, and 12 g Catalyst.
  • Thin sections (.5-2.0 micron) with excellent morphological structure preservation
  • Water-clear blocks; casts in 90 minutes max. at room temp
  • Good lipid and enzyme retention when processing at low temperatures (4°C)
  • Removal of JB-4 resin prior to staining is not necessary
  • Clearing agents such as xylene and chloroform are not needed
  • Higher clarity and contrast than with paraffin sections
  • Easy processing of difficult specimens such as calcified bone and delicate embryonic tissue
  • It produces harder blocks which is ideal for dense samples, such as bone
Product Family Options
  • Désignation
    Taille
  • JB-4 Solution B
    30 mL
    Each (30ml)
    Each (30ml) - $137.66
    $137.66
    Each (30ml)
       $137.66
     : 14270-04
  • JB-4 Embedding Kit
    Kit
    Each
    Each - $1,323.68
    $1,323.68
    Kit of 1
    Kit of 1 - $1,323.68
    $1,323.68
    Each
       $1,323.68
    Kit of 1
       $1,323.68
     : 14270-00