Inoculating Loops, Electron Microscopy Sciences
Our high quality inoculating loops are made from platinum wire. The 15% iridium adds necessary rigidity permitting thinner wire for delicate work. The loop section is welded and smoothly finished to form a “perfect circle”.
Our high quality inoculating loops are made from platinum wire. The 15% iridium adds necessary rigidity permitting thinner wire for delicate work. The loop section is welded and smoothly finished to form a “perfect circle”.
Product Family Options
- DésignationTailleJauge
- Inoculating Loops3 x 45 mm24 Gauge-: -
- : 62436-03
- : 62433-05
- : 62436-05
- : 62434-02
- : 62437-02
- : 62434-03
- : 62437-03
- : 62437-05