Inoculating Loop/Needle, Electron Microscopy Sciences
Inoculating Loop
  100489-540
 :  72952-10
Inoculating Loop/Needle, Electron Microscopy Sciences
  100489-540
Inoculating Loop/Needle, Electron Microscopy Sciences
Inoculating Loop/Needle, Electron Microscopy Sciences
Inoculating Loop
$65.33
Each Pack of 12
Nichrome wire. An 8” aluminum handle with a 3” nichrome wire, 25 gauge. Loop O.D. is 4 mm; loop I.D. is 2.5 mm. Available with a loop or a needle.
  • Désignation:
    Inoculating Loop