Histology Containers, Electron Microscopy Sciences
À propos de cet article
The 20 m to 1 L sizes are supplied with tight-fitting screw caps and warning labels for 10% Formalin with extra space for writing
- Designed for storage and transport of histology specimens
- Made from durable polypropylene (except 8L size which is made from polyethylene).
- Leak proof seal caps.
The 2 to 8L sizes are supplied with snap-top lids and carrying handles, and has no graduations.
All of the containers (except the 5 mL size) are graduated on three sides.
9 Options Available Below
Product Details & Documents
The 20 m to 1 L sizes are supplied with tight-fitting screw caps and warning labels for 10% Formalin with extra space for writing
- Designed for storage and transport of histology specimens
- Made from durable polypropylene (except 8L size which is made from polyethylene).
- Leak proof seal caps.
The 2 to 8L sizes are supplied with snap-top lids and carrying handles, and has no graduations.
All of the containers (except the 5 mL size) are graduated on three sides.
Product Family Options
Informations sur le produit
- CapacitéColorAvailabilityPrice
- 60 mlSemi-ClearEach$114.50Pack of 100$87.77Supplier Number: 64233-14
- 5LWhiteEach$484.60Pack of 10$369.56Supplier Number: 64233-26
- 5 mlSemi-ClearDiscontinued-Supplier Number: -
- 20 mlSemi-ClearEach$377.61Pack of 200$287.92Supplier Number: 64233-11
- 90 mlSemi-ClearEach$484.15Pack of 400$369.17Supplier Number: 64233-16
- 120 mlSemi-ClearEach$929.71Pack of 300$709.39Supplier Number: 64233-17
- 500 mlOpaqueEach$500.24Pack of 100$381.76Supplier Number: 64233-20
- Discontinued-Supplier Number: -
- OpaqueEach$557.46Pack of 100$424.72Supplier Number: 64233-22
- 40 mlSemi-ClearEach$196.79Pack of 200$149.71Supplier Number: 64233-12
- 2 LWhiteEach$525.57Pack of 20$401.21Supplier Number: 64233-24