High-Intensity UV Lamp, Electron Microscopy Sciences
About this item
These UV Lamps are designed to provide a beam of 365 nm wave with an average 7000 uW/cm² at 38.2 cm (15"). Its powerful 100 watt light source fluoresces fine particles and hard to see surface defects. The lifetime of the light bulb is 2000 hours.
At 45.7 cm (18"), this lamp produces a highly intense center of UV irradiance about 12.5 cm (5") in diameter. This offers the microscopist a quality tool in staining and polymerization as well as many other uses that are related to UV lights in EM labs. The lamp head rotates a full 3600, and is mounted into a removable pistol grip handle, which inserts firmly into the transformer base (as a stand); it weighs only 2.5 lbs. It comes with a 12.5 cm (5") diameter black light filter which is recessed in the head. If more details are needed in order to study the subject, a second filter is available as an option that reduces the visible light. Powered by 115 V, 60 Hz, total cord length is 4.57 m (15 ft.).
2 Options Available Below
Product Details & Documents
These UV Lamps are designed to provide a beam of 365 nm wave with an average 7000 uW/cm² at 38.2 cm (15"). Its powerful 100 watt light source fluoresces fine particles and hard to see surface defects. The lifetime of the light bulb is 2000 hours.
At 45.7 cm (18"), this lamp produces a highly intense center of UV irradiance about 12.5 cm (5") in diameter. This offers the microscopist a quality tool in staining and polymerization as well as many other uses that are related to UV lights in EM labs. The lamp head rotates a full 3600, and is mounted into a removable pistol grip handle, which inserts firmly into the transformer base (as a stand); it weighs only 2.5 lbs. It comes with a 12.5 cm (5") diameter black light filter which is recessed in the head. If more details are needed in order to study the subject, a second filter is available as an option that reduces the visible light. Powered by 115 V, 60 Hz, total cord length is 4.57 m (15 ft.).
Product Family Options
Product Information