High-Density Polyethylene Staining Jar, Electron Microscopy Sciences
Complete Unit, 4 1/2 x 2 1/2"
Catalog # 100500-234
 :  70319
High-Density Polyethylene Staining Jar, Electron Microscopy Sciences
Catalog # 100500-234
High-Density Polyethylene Staining Jar, Electron Microscopy Sciences
High-Density Polyethylene Staining Jar, Electron Microscopy Sciences
Complete Unit, 4 1/2 x 2 1/2"
Price
$114.01
Log in pour la disponibilité
Case of 4 Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
An unbreakable, high-density polyethylene staining jar with internal grooves and a shallow screw cap.
The jar holds 5 single 1 x 3" slides or 10 back-to-back slides vertically.
Overall measurement H x Dia: 4 1/2 x 2 1/2".
Spécifications
  • Désignation:
    Complete Unit
  • Dimensions:
    4 1/2 x 2 1/2"