High Precision Diamond Scribing Tools, Electron Microscopy Sciences
High Precision Diamond Scribing Tools, Electron Microscopy Sciences
High Precision Diamond Scribing Tools, Electron Microscopy Sciences
Certificates
High Precision Diamond Scribing Tools, Electron Microscopy Sciences
High Precision Diamond Scribing Tools, Electron Microscopy Sciences
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A newly designed diamond scribing tool set containing eight precision natural diamond scribes in various sizes and configurations. This set is perfect for precision work in microscopy. The set consists of eight diamond tips, 3 mm in diameter and are mounted in interchangeable tool cones and an MT-1 tool handle. The tool and handle are all housed in a hardwood instrument case. Individual scribers can be purchased separately.
Product Family Options
Product Information
  • Désignation
    Diameter
    Longueur
    Availability
    Price
  • High precision diamond scriber, bent tip
    0.01" tip
    0.006" tip length
    Log in for availability
    Log in for availability
    Each
    Each - $257.47
    $257.47
    Each
       $257.47
    Supplier Number: 62105-B2
  • High precision diamond scriber, straight tip
    0.01" tip
    0.006" tip length
    Log in for availability
    Log in for availability
    Each
    Each - $216.64
    $216.64
    Each
       $216.64
    Supplier Number: 62105-S2
  • High precision diamond scriber, bent tip
    0.02" tip
    0.006" tip length
    Discontinued
    Supplier Number: 62105-B1
  • High precision diamond scriber, straight tip
    0.03" tip
    0.006" tip length
    Log in for availability
    Log in for availability
    Each
    Each - $182.55
    $182.55
    Each
       $182.55
    Supplier Number: 62105-S1
  • High precision diamond scriber, straight heavy-duty tip
    0.02" tip
    0.12" tip length
    Discontinued
    Supplier Number: 62105-HS
  • High precision diamond scriber
    0.02" tip
    0.2" tip length
    Log in for availability
    Log in for availability
    Each
    Each - $240.39
    $240.39
    Each
       $240.39
    Supplier Number: 62105-BE
  • High precision diamond scriber
    0.03" tip
    0.12" tip length
    Discontinued
    Supplier Number: 62105-HC
  • High precision diamond scriber, set
    Log in for availability
    Log in for availability
    Each
    Each - $1,515.62
    $1,515.62
    Each
       $1,515.62
    Supplier Number: 62105-00