Hex Key Wrenches, Electron Microscopy Sciences | Avantor
Hex Key Wrenches, Electron Microscopy Sciences
Hex Key Wrenches, Electron Microscopy Sciences
Certificates
Hex Key Wrenches, Electron Microscopy Sciences
Hex Key Wrenches, Electron Microscopy Sciences
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from heat treated chrome nickel steel. Useful to have around the lab. Its sturdy handle, folds up just like a pocket knife. Can be used at any angle.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 100497-570
  • 100497-564
  • 100497-568
  • 100497-566
  • Type:
  • Hex Key
  • Hex Key
  • Hex Key
  • Hex Key
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Hex Key, Metric Set 2
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $66.47
    $66.47
    Each
       $66.47
    Supplier Number: 62076-02
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100497-570
    Type
    Hex Key
    Minority or Woman-Owned Business Enterprise
  • Hex Key, English Set 1
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $41.37
    $41.37
    Each
       $41.37
    Supplier Number: 62075-01
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100497-564
    Type
    Hex Key
    Minority or Woman-Owned Business Enterprise
  • Hex Key, Metric Set 1
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $48.21
    $48.21
    Each
       $48.21
    Supplier Number: 62076-01
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100497-568
    Type
    Hex Key
    Minority or Woman-Owned Business Enterprise
  • Hex Key, English Set 2
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $55.59
    $55.59
    Each
       $55.59
    Supplier Number: 62075-03
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100497-566
    Type
    Hex Key
    Minority or Woman-Owned Business Enterprise