Hand Piece Replacement, Electron Microscopy Sciences
Hand Piece Replacement, Electron Microscopy Sciences
Hand Piece, Procraft
Catalog # 100497-906
Supplier #:  62137-01
Hand Piece Replacement, Electron Microscopy Sciences
Catalog # 100497-906
Hand Piece Replacement, Electron Microscopy Sciences
Hand Piece Replacement, Electron Microscopy Sciences
Hand Piece, Procraft
Price
$103.38
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Hand Piece with the chuck takes accessory shanks from 0 to 4 mm (0 to 5/32").
Spécifications
  • Désignation:
    Hand Piece, Procraft