H.S.E. Test Slide for Phase Contrast Microscopy, Electron Microscopy Sciences
Product Details & Documents
Epoxy replica of a master slide. The replicas are mounted on microscope slides of 1.2 mm thickness with cover glass of 0.17 mm thickness.
This information is transferred to the accompanying table, which indicates the maximum phase change passing through test objects to test slide. A satisfactory system will detect block 5.
Certifications: This test slide is made in the UK and is under license certification from the National Physical Laboratory.
Delivery information: Full details are supplied with the slide.
Spécifications
- Désignation:Hse test slides (s84) for calibration in asbestos analysis