Grided Microscope Slides without hydrophobic coating, Electron Microscopy Sciences
Grided Microscope Slides without hydrophobic coating, Electron Microscopy Sciences
Grided Microscope Slides without hydrophobic coating, Electron Microscopy Sciences
Grided Microscope Slides without hydrophobic coating, Electron Microscopy Sciences
Grided Microscope Slides without hydrophobic coating, Electron Microscopy Sciences
These 3×1" slides have an indexed grid on the reverse side of the frosted end. The ink is resistant to stains and common laboratory solvents.
  • Each square grid fills one low-powered-field (approximately 100× magnification)
  • Easy to locate an organism or cell every time
  • Eighty one squares
The slides are available with coatings of Poly-L-Lysine or Silane, or they can be purchased plain (uncoated). The grid measures 2 mm square, 16 mm total. Numbers 1-8 and alpha A-H.
Product Family Options
  • Désignation
  • Grid slides, round well
    Each
    Each - $326.23
    $326.23
    Pack of 72
    Pack of 72 - $233.29
    $233.29
    Each
       $326.23
    Pack of 72
       $233.29
     : 63408-01
  • Grid slides, poly-l-lysine
    Each
    Each - $253.66
    $253.66
    Pack of 72
    Pack of 72 - $230.13
    $230.13
    Each
       $253.66
    Pack of 72
       $230.13
     : 63406-01
  • Grid slides, poly-l-lysine
    Each
    Each - $482.79
    $482.79
    Pack of 144
    Pack of 144 - $349.94
    $349.94
    Each
       $482.79
    Pack of 144
       $349.94
     : 63406-02
  • Grid slides, uncoated
    Each
    Each - $226.37
    $226.37
    Pack of 72
    Pack of 72 - $205.37
    $205.37
    Each
       $226.37
    Pack of 72
       $205.37
     : 63405-01
  • Grid slides, uncoated
    Each
    Each - $440.27
    $440.27
    Pack of 144
    Pack of 144 - $330.90
    $330.90
    Each
       $440.27
    Pack of 144
       $330.90
     : 63405-02
  • Grid slides, square well
    Each
    Each - $326.23
    $326.23
    Pack of 72
    Pack of 72 - $233.29
    $233.29
    Each
       $326.23
    Pack of 72
       $233.29
     : 63409-02