Grid Storage Box, 50 Capacity, Electron Microscopy Sciences
À propos de cet article
Storage for 50 grids in deep diamond-shaped wells. All wells are identified. The base is resistant to organics and reactions can be carried out on grid-mounted samples in the wells. Complete with grid recording card.
2 Options Available Below
Product Details & Documents
Storage for 50 grids in deep diamond-shaped wells. All wells are identified. The base is resistant to organics and reactions can be carried out on grid-mounted samples in the wells. Complete with grid recording card.
Product Family Options
Informations sur le produit
- DésignationCapacitéAvailabilityPrice
- Supplier Number: 71150
- Grid Storage Box, Clear Cover50 GridsDozen$177.22Each$177.22Supplier Number: 71152