Grid Mats, Electron Microscopy Sciences | Avantor
Grid Mats, Electron Microscopy Sciences
Grid Mats, Electron Microscopy Sciences
Certificates
Grid Mats, Electron Microscopy Sciences
Grid Mats, Electron Microscopy Sciences
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
White silicone rubber mats, with numbered compartments.
  • Good for organizing grids
  • They will not slide or jump between compartments
  • Ideal for staining grids
Easy to pick up grids without damaging forcep tips. Mats are available for square and round petri dishes, (100 mm diameter).
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Round grid mats
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Dozen
    Dozen - $245.00
    $245.00
    Log in pour la disponibilité
    Each
    Each - $245.00
    $245.00
    Dozen
       $245.00
    Each
       $245.00
    Supplier Number: 71172
  • Round grid mats
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $24.14
    $24.14
    Each
       $24.14
    Supplier Number: 71170
  • Square grid mats
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Dozen
    Dozen - $219.34
    $219.34
    Log in pour la disponibilité
    Each
    Each - $219.34
    $219.34
    Dozen
       $219.34
    Each
       $219.34
    Supplier Number: 71162
  • Square grid mats
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $20.87
    $20.87
    Each
       $20.87
    Supplier Number: 71160